Key Documents
Z107476
Sigma-Aldrich
Aldrich® fritted adapter
female joint: ST/NS 24/40, male joint: ST/NS 24/40
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Joint
female joint: ST/NS24/40
male joint: ST/NS24/40
Related Categories
General description
The extra-coarse fritted disc (diameter: 20 mm, porosity A, 145-174 micron) prevents solids from being pulled into the condenser when removing solvents from solid-solvent mixtures. Overall adapter length: 150 mm.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)