Skip to Content
Merck
All Photos(3)

Documents

W530597

Sigma-Aldrich

Sabinene

greener alternative

natural, 75%

Synonym(s):

4(10)-thujene, Sabinen, 4-methylidene-1-propan-2-ylbicyclo[3.1.0]hexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
EC Number:
Council of Europe no.:
11018
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
1.059
NACRES:
NA.21

grade

Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

Assay

75%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

density

0.842 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

woody; citrus; spicy

SMILES string

CC(C)C12CCC(=C)C1C2

InChI

1S/C10H16/c1-7(2)10-5-4-8(3)9(10)6-10/h7,9H,3-6H2,1-2H3

InChI key

NDVASEGYNIMXJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is Biobased and thus aligns with "Less Hazardous Chemical Syntheses" and "Use of Renewable Feedstocks".

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamed A Ibrahim et al.
Molecules (Basel, Switzerland), 25(3) (2020-02-06)
The intent of this study was to utilize distillation timeframes (DT) of nutmeg (Myristica fragrans) essential oil (EO) to generate fractions with differential chemical compositions and bioactivity. Ten fractions were captured at the following distillation timeframes: 0.0-0.5, 0.5-1.0, 1.0-2.5, 2.5-5.0
L Chiappini et al.
Analytical and bioanalytical chemistry, 386(6), 1749-1759 (2006-09-15)
A new one-step method for the analysis of highly polar components of secondary organic aerosols (SOA) has been developed. This method should lead to a better understanding of SOA formation and evolution since it enables the compounds responsible for SOA
Lars Müller et al.
Rapid communications in mass spectrometry : RCM, 23(7), 971-979 (2009-02-27)
The components of secondary organic aerosols (SOAs) generated from the gas-phase ozonolysis of two C(10)H(16)-terpenes (alpha-pinene; sabinene) and a cyclic C(6)H(10) alkene (cyclohexene) were characterized by the use of a Fourier transform ion cyclotron mass spectrometer equipped with an electrospray
José Joaquín Merino et al.
Journal of clinical medicine, 8(2) (2019-02-03)
Introduction: Bruxism affects teeth and provokes sleep alterations. We evaluated whether saliva Myeloperoxidase (MPO) activity could be reduced after 21 neurofeedback training (NO) sessions in Origanum majorana (AE) bruxistic-treated patients (n = 12 patients, 120 saliva samples). The term divergence
Rubén Olmedo et al.
Journal of food science and technology, 56(5), 2522-2535 (2019-06-07)
This study compared the antioxidant properties of oregano essential oil (OEO) and butylated hydroxytoluene (BHT) alone and when combined. The principal components in OEO were gamma terpinene (25.1 g/100 g), terpinen-4-ol (16.7 g/100 g), and carvacrol (16.2 g/100 g). OEO showed 60% DPPH inhibition and 10 mg/g

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service