Skip to Content
Merck
All Photos(1)

Key Documents

W505005

Sigma-Aldrich

Diethyl maleate

≥96%, FG

Synonym(s):

Maleic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCOCH=CHCOOC2H5
CAS Number:
Molecular Weight:
172.18
Beilstein:
1100825
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.351
NACRES:
NA.21
grade:
FG
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

FG

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

vapor density

5.93 (vs air)

vapor pressure

1 mmHg ( 14 °C)

Assay

≥96%

autoignition temp.

662 °F

refractive index

n20/D 1.441 (lit.)

bp

225 °C (lit.)

mp

−10 °C (lit.)

density

1.064 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

banana; fruity; citrus

SMILES string

CCOC(=O)\C=C/C(=O)OCC

InChI

1S/C8H12O4/c1-3-11-7(9)5-6-8(10)12-4-2/h5-6H,3-4H2,1-2H3/b6-5-

InChI key

IEPRKVQEAMIZSS-WAYWQWQTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl maleate is reported to occur in the cabernet sauvignon wine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Some volatile components of Vitis vinifera variety Cabernet Sauvignon wine.
Slingsby RW, et al.
American Journal of Enology and Viticulture, 31(4), 360-363 (1980)
Yosuke Hirotsu et al.
Nucleic acids research, 40(20), 10228-10239 (2012-09-12)
NF-E2-related factor 2 (Nrf2) is a key transcription factor that is critical for cellular defense against oxidative and xenobiotic insults. Nrf2 heterodimerizes with small Maf (sMaf) proteins and binds to antioxidant response elements (AREs) to activate a battery of cytoprotective
Nazir M Khan et al.
Free radical biology & medicine, 51(1), 115-128 (2011-05-03)
There are no reports describing the ability of pro-oxidants to protect against radiation-induced apoptosis. Activation of the redox-sensitive transcription factor Nrf2 by low levels of ROS is known to protect against oxidative stress-induced cell death. In this study, hydrogen peroxide
Lauren Drowley et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 18(10), 1865-1873 (2010-07-29)
Although cellular transplantation has been shown to promote improvements in cardiac function following injury, poor cell survival following transplantation continues to limit the efficacy of this therapy. We have previously observed that transplantation of muscle-derived stem cells (MDSCs) improves cardiac
Nobuhiko Harada et al.
Archives of biochemistry and biophysics, 508(1), 101-109 (2011-02-10)
Iron is an essential element of hemoglobin, and efficient iron recycling from senescent erythrocytes by splenic macrophages is required for erythrocyte hemoglobin synthesis during erythropoiesis. Ferroportin 1 (Fpn1) is the sole iron exporter in mammals, and it also regulates iron

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service