Skip to Content
Merck
All Photos(1)

Key Documents

W444165

Sigma-Aldrich

Jasmolactone

≥95%

Synonym(s):

Petal pyranone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O2
CAS Number:
Molecular Weight:
168.23
FEMA Number:
4441
MDL number:
UNSPSC Code:
12164502
Flavis number:
10.040
NACRES:
NA.21
grade:
Fragrance grade
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

Fragrance grade

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009

Assay

≥95%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

apricot; coconut; jasmine; fruity; floral; peach

SMILES string

O1C(CCCC1=O)CC\C=C\C

InChI

1S/C10H16O2/c1-2-3-4-6-9-7-5-8-10(11)12-9/h2-3,9H,4-8H2,1H3/b3-2+

InChI key

NBCMACYORPIYNY-NSCUHMNNSA-N

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service