Skip to Content
Merck
All Photos(1)

Key Documents

W401501

Sigma-Aldrich

Pyrazine

≥99%, FG

Synonym(s):

1,4-diazabenzene, 1,4-diazine, p-diazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2
CAS Number:
Molecular Weight:
80.09
FEMA Number:
4015
Beilstein:
103905
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.144
NACRES:
NA.21
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥99%

bp

115-116 °C (lit.)

mp

50-56 °C (lit.)

density

1.031 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

pungent; nutty

SMILES string

c1cnccn1

InChI

1S/C4H4N2/c1-2-6-4-3-5-1/h1-4H

InChI key

KYQCOXFCLRTKLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily J Dennis et al.
Nature, 562(7725), 119-123 (2018-09-28)
DEET (N,N-diethyl-meta-toluamide) is a synthetic chemical identified by the US Department of Agriculture in 1946 in a screen for repellents to protect soldiers from mosquito-borne diseases1,2. Since its discovery, DEET has become the world's most widely used arthropod repellent and
Nicole Favreau-Farhadi et al.
Journal of food science, 80(10), C2140-C2146 (2015-09-27)
Rosmarinic acid and Epigallocatechin gallate concentrations were studied as natural inhibitors of Maillard browning in glucose/glycine model systems, and in bakery rolls and applesauce. The concentrations of the inhibitors were varied to determine the highest level of inhibition without a
Gustavo Luis Leonardo Scalone et al.
Journal of agricultural and food chemistry, 63(22), 5364-5372 (2015-05-15)
Pyrazines are specific Maillard reaction compounds known to contribute to the unique aroma of many products. Most studies concerning the generation of pyrazines in the Maillard reaction have focused on amino acids, while little information is available on the impact

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service