Skip to Content
Merck
All Photos(1)

Key Documents

W379301

Sigma-Aldrich

D-(−)-Ribose

≥98%

Synonym(s):

Dextro-ribose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
FEMA Number:
3793
Beilstein:
1723081
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21
Organoleptic:
odorless
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

reg. compliance

FDA 21 CFR 117

Assay

≥98%

optical activity

[α]21/D −19.7°, c = 4 in H2O

mp

88-92 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

OC[C@@H](O)[C@@H](O)[C@@H](O)C([H])=O

InChI

1S/C5H10O5/c6-1-3(8)5(10)4(9)2-7/h1,3-5,7-10H,2H2/t3-,4+,5-/m0/s1

InChI key

PYMYPHUHKUWMLA-LMVFSUKVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-(−)-Ribose has been in an in vitro study of the effect of non-enzymatic ribation in modifying bone collagen leading to bone fragility.

Application

  • Protective effect of thymoquinone on glycation of human myoglobin induced by d-ribose.: This study investigates the protective effects of thymoquinone against d-ribose-induced glycation in human myoglobin, highlighting potential therapeutic applications in preventing protein glycation (Liu et al., 2023).

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D-ribose and deoxy-D-ribose induce apoptosis in human quiescent peripheral blood mononuclear cells.
Barbieri D, et al.
Biochemical and Biophysical Research Communications, 201(3), 1109-1116 (1994)
Alice Pavlowsky et al.
Current biology : CB, 28(11), 1783-1793 (2018-05-22)
Memory consolidation is a crucial step for long-term memory (LTM) storage. However, we still lack a clear picture of how memory consolidation is regulated at the neuronal circuit level. Here, we took advantage of the well-described anatomy of the Drosophila
Raman K Sharma et al.
Bioorganic & medicinal chemistry, 20(23), 6821-6830 (2012-10-27)
A series of peracetylated O-aryl α,β-d-ribofuranosides have been synthesized and an efficient biocatalytic methodology has been developed for the separation of their anomers which was otherwise almost impossible by column chromatographic or other techniques. The incubation of 2,3,5-tri-O-acetyl-1-O-aryl-α,β-d-ribofuranoside with Lipozyme®
Carine Baraguey et al.
Organic & biomolecular chemistry, 11(16), 2638-2647 (2013-03-05)
The pivaloyloxymethyl (PivOM) group is a biolabile 2'-O-ribose protection that is under development in a prodrug-based approach for siRNA applications. Besides an expected cellular uptake, nucleic acid sequences carrying PivOM showed also increased nuclease resistance and, in most cases, an
Xiang-Guo Li et al.
Chemical communications (Cambridge, England), 49(35), 3682-3684 (2013-03-29)
Peptide glycosylation with 5-deoxy-5-[(18)F]fluororibose was translated into preclinical settings. The novel (18)F-labeled Siglec-9 peptide was produced using an automated synthesis procedure. The (18)F-labeled Siglec-9 peptide showed favorable binding in the animal model of inflammation in vivo.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service