Skip to Content
Merck
All Photos(1)

Key Documents

W362301

Sigma-Aldrich

5-Ethyl-4-hydroxy-2-methyl-3(2H)-furanone, mixture of isomers

96%, FG

Synonym(s):

Homofuraneol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O3
CAS Number:
Molecular Weight:
142.15
FEMA Number:
3623
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.084
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

96%

refractive index

n20/D 1.512 (lit.)

bp

248-249 °C (lit.)

density

1.137 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

bread; burnt; caramel; maple; sweet

SMILES string

CCC1=C(O)C(C(C)O1)=O

InChI

1S/C7H10O3/c1-3-5-7(9)6(8)4(2)10-5/h4,9H,3H2,1-2H3

InChI key

QJYOEDXNPLUUAR-UHFFFAOYSA-N

Related Categories

General description

5-Ethyl-4-hydroxy-2-methyl-3(2H)-furanone is one of the key volatile components of shoyu (soy sauce). It also occurs in roasted coffee and blackberries.

Application

5-Ethyl-4-hydroxy-2-methyl-3(2H)-furanone (a mixture of isomers) can be used as a flavoring agent in food industries due to its excellent sensory properties.

Packaging

Packaged in glass bottles

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Aroma simulation on the basis of the odourant composition of roasted coffee headspace
Mayer F & Grosch W.
Flavour and Fragrance Journal, 16(3), 180-190 (2001)
Aroma extract dilution analysis of cv. Marion (Rubus spp. hyb) and cv. Evergreen (R. laciniatus L.) blackberries
Klesk K & Qian M.
Journal of Agricultural and Food Chemistry, 51(11), 3436-3441 (2003)
X Li et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 36(4), 305-314 (1998-07-03)
Components with DNA breaking activity in soy sauce were investigated. It was found that there were water soluble high molecular weight DNA breaking components in soy sauce. Two DNA breaking components in the ethyl acetate extract of soy sauce were
K Hiramoto et al.
Mutation research, 415(1-2), 79-83 (1998-08-26)
Fragrant hydroxyfuranone and dihydroxypyranone derivatives generated in the Maillard reaction of sugars and amino acids are detected in various processed foods and have been shown active to break DNA single-strand in the in vitro studies. In the present study, absorption
T Sasaki et al.
Bioscience, biotechnology, and biochemistry, 62(10), 1865-1869 (1998-12-04)
We determined the anti-cataract effects and antioxidative activities of four 4-hydroxy-3(2H)-furanones. These four furanones showed similar antioxidative activities in the ferric ion reduction model. 4-Hydroxy-2,5-dimethyl-3(2H)-furanone (HDMF) and 2(or 5)-ethyl-4-hydroxy-5(or 2)-methyl-3(2H)-furanone (EHMF) exhibited a higher suppression effect on lipid peroxidation in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service