Skip to Content
Merck
All Photos(2)

Key Documents

W355901

Sigma-Aldrich

γ-Terpinene

natural, 95%, FG

Synonym(s):

1-Methyl-4-(1-methylethyl)-1,4-cyclohexadiene, 1-Isopropyl-4-methyl-1,4-cyclohexadiene, p-Mentha-1,4-diene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
FEMA Number:
3559
Beilstein:
2038347
EC Number:
Council of Europe no.:
11025
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
1.020
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 173.65 (172.515)

vapor density

4.7 (vs air)

vapor pressure

~0.7 mmHg ( 20 °C)

Assay

95%

refractive index

n20/D 1.474 (lit.)

bp

182 °C (lit.)

density

0.85 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

herbaceous; citrus

SMILES string

CC(C)C1=CCC(C)=CC1

InChI

1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4,7-8H,5-6H2,1-3H3

InChI key

YKFLAYDHMOASIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3 - Repr. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Wei-bin Yuan et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 34(7), 1067-1069 (2011-11-10)
To analysis the constituents of volatile oil from Fructus Auranti Immaturus by GC-MS. The volatile oil was extracted by steam distillation, then separated by capillary gas chromatography. The constituents of volatile oil were identified and their amount were determined by
Nabila Bouderdara et al.
Natural product communications, 6(1), 115-117 (2011-03-04)
The essential oil of aerial parts of Cachrys libanotis L. (Apiaceae) from east Algeria was extracted by hydrodistillation and analyzed by GC-FID and GC-MS. Thirty-one compounds were identified, the main components being germacrene-D (18.0%), gamma-terpinene (6.4%), p-cymene (5.5%), caryophyllene oxide
Amr E Edris
Journal of dietary supplements, 8(1), 34-42 (2012-03-22)
The crude oil of Nigella sativa L. (Black cumin) has well-known nutraceutical and pharmaceutical properties. The oil is prone to rapid oxidative deterioration because of its high content of poly-unsaturated fatty acids. In the current investigation, different cold-pressed crude oils
Nataraj Jagannath et al.
Natural product communications, 7(7), 943-946 (2012-08-23)
The essential oil was extracted from the seeds of Heracleum rigens by hydrodistillation and a total of twenty compounds accounting for 98.5% of the total oil composition were identified. Physicochemical properties and chemical composition of the oil was determined by
Xiao-Dong Li et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 34(10), 1544-1548 (2012-03-01)
To extract the essential oils from the Seedlings, the Aseptic Seedlings and the Tissue Culture Seedlings of Thymus vulgaris and analyze their chemical components and the relative contents. The essential oils were extracted by steam distillation, the chemical components and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service