Skip to Content
Merck
All Photos(1)

Key Documents

W351504

Sigma-Aldrich

1-Octen-3-one

50 wt. % in 1-octen-3-ol, stabilized, FG

Synonym(s):

N-pentyl vinyl ketone, Vinyl amyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
3515
Council of Europe no.:
2312
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.081
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002

may contain

~0.10% α-Tocopherol as stabilizer

concentration

50 wt. % in 1-octen-3-ol

refractive index

n20/D 1.4359

bp

174-182 °C

density

0.843 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

mushroom; herbaceous; earthy

SMILES string

O=C(CCCCC)C=C

InChI

1S/C8H14O/c1-3-5-6-7-8(9)4-2/h4H,2-3,5-7H2,1H3

InChI key

KLTVSWGXIAYTHO-UHFFFAOYSA-N

Related Categories

General description

1-Octen-3-one is one of the main volatile flavor compounds of mushroom. It has also been reported to play a major in the resistance of Trifolium subterraneum cotyledons against damage caused by red-legged earth mite.

Packaging

Packaged in glass bottles

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

145.0 °F - closed cup

Flash Point(C)

62.78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Correlation of 1-octen-3-one with antixenotic resistance in subterranean clover cotyledons to red-legged earth mite, Halotydeus destructor (Acarina: Penthaleidae).
Jiang Y, et al.
Journal of Chemical Ecology, 22(2), 369-382 (1996)
Studies on the enzymic reduction of 1-octen-3-one in mushroom (Agaricus bisporus).
Chen CC & W CM.
Journal of Agricultural and Food Chemistry, 32(6), 1342-1344 (1984)
Meryl B Lubran et al.
Journal of agricultural and food chemistry, 53(21), 8325-8327 (2005-10-13)
Taste threshold tests of ferrous sulfate (FeSO4) solutions have been confounded by the presence of putative odorants. To detect the presence of odorants released from these solutions solid-phase microextraction (SPME) was used to collect volatiles in the headspace above FeSO4
M A Drake et al.
Journal of dairy science, 93(11), 5069-5081 (2010-10-23)
A current industry goal is to produce a 75 to 80% fat-reduced Cheddar cheese that is tasty and appealing to consumers. Despite previous studies on reduced-fat cheese, information is critically lacking in understanding the flavor and flavor chemistry of reduced-fat
Volatile constituents of mushrooms (Agaricus bisporus). Changes which occur during heating.
Picardi SM & Issenberg P.
Journal of Agricultural and Food Chemistry, 21(6), 959-962 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service