Skip to Content
Merck
All Photos(1)

Key Documents

W350702

Sigma-Aldrich

Isoamyl isobutyrate

≥98%, FG

Synonym(s):

3-methylbutyl 2-methylpropanoate, Isopentyl isobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2O2CCH(CH3)2
CAS Number:
Molecular Weight:
158.24
FEMA Number:
3507
EC Number:
Council of Europe no.:
294
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.419
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.406 (lit.)

bp

169-171 °C (lit.)

density

0.856 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; waxy; fruity; sweet

SMILES string

CC(C)CCOC(=O)C(C)C

InChI

1S/C9H18O2/c1-7(2)5-6-11-9(10)8(3)4/h7-8H,5-6H2,1-4H3

InChI key

VFTGLSWXJMRZNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Isoamyl isobutyrate has been reported to be one of the hop aroma constituents in beer.[1]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hop flavor constituents in beer by headspace analysis.
Murakami A, et al.
Zeitschrift fur Lebensmittel-Untersuchung Und -Forschung, 45(1) (1987)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service