Skip to Content
Merck
All Photos(1)

Key Documents

W342408

Sigma-Aldrich

3-Acetylpyridine

≥98%, FG

Synonym(s):

Methyl 3-pyridyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO
CAS Number:
Molecular Weight:
121.14
FEMA Number:
3424
Beilstein:
107751
EC Number:
Council of Europe no.:
2316
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.039
NACRES:
NA.21
Organoleptic:
nutty; sweet
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥98%

impurities

≤0.5% Water (Karl Fischer)

refractive index

n20/D 1.534 (lit.)

bp

220 °C (lit.)

mp

11-13 °C (lit.)

density

1.102 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

nutty; sweet

SMILES string

CC(=O)c1cccnc1

InChI

1S/C7H7NO/c1-6(9)7-3-2-4-8-5-7/h2-5H,1H3

InChI key

WEGYGNROSJDEIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Identification of antioxidant and flavour marker compounds in Kalosi-Enrekang Arabica brewed coffee processed using different postharvest treatment methods.: This study explores the impact of various postharvest processes on the antioxidant properties and flavor profile of Kalosi-Enrekang Arabica coffee, with a focus on 3-acetylpyridine as a key flavor marker. The research aims to enhance coffee quality and consumer satisfaction through optimized processing techniques (Yulianti et al., 2024).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ai-Li Cai et al.
The European journal of neuroscience, 24(8), 2169-2176 (2006-10-18)
Zinc neurotoxicity has been demonstrated in ischemic, seizure, hypoglycemic, and trauma-induced neuronal death where Zn(2+) is thought to be synaptically released and taken up in neighbouring neurons, reaching toxic concentrations. We previously demonstrated that toxicity of extracellular Zn(2+) depended on
T M Kuchmerovskaia et al.
Voprosy meditsinskoi khimii, 48(3), 264-270 (2002-09-24)
The effect of administration of 3-acetylpyridine (antivitamin B3), on rat brain serotoninergic and NAD receptor systems was investigated. The injection of 3-acetylpyridine to rats caused a decrease of NAD and serotonin content in the brain and changes in [U-14C]NAD binding
Amanda Sierra et al.
Endocrine, 21(1), 43-51 (2003-06-05)
The expression of aromatase, the enzyme that transforms testosterone into estradiol, was analyzed by reverse transcriptase polymerase chain reaction in the inferior olive of adult male rats. The expression of this messenger in the inferior olive suggests that this brain
Mahyar Janahmadi et al.
Neurotoxicology, 30(3), 393-402 (2009-05-16)
Riluzole has been shown to possess neuroprotective effects in a variety of neurological and animal model of diseases, including motor diseases. However, the mechanism(s) by which riluzole preserves the intrinsic electrophysiological characteristics of neuronal membrane has not been fully delineated.
Sheldon J Moss et al.
Neuroscience letters, 321(1-2), 53-56 (2002-03-02)
There are no published reports assessing whether the non-immunosuppressant immunophilin ligand GPI-1046, which has putative neurotrophic effects in a variety of models of neurotoxicity, possesses neuroprotective effects in the 3-acetylpyridine (3AP) toxicity model in rats. The purpose of the present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service