Skip to Content
Merck
All Photos(1)

Key Documents

W331902

Sigma-Aldrich

L-Proline

99%, FCC, FG

Synonym(s):

(S)-Pyrrolidine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
FEMA Number:
3318
Beilstein:
80810
EC Number:
Council of Europe no.:
10378
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
17.019
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FCC
FDA 21 CFR 172.320

Assay

99%

optical activity

[α]20/D −84°, c = 4 in H2O

mp

228 °C (dec.) (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

OC(=O)[C@@H]1CCCN1

InChI

1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)/t4-/m0/s1

InChI key

ONIBWKKTOPOVIA-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie Couturier et al.
The Journal of biological chemistry, 288(20), 14624-14635 (2013-04-06)
The microbial deconstruction of the plant cell wall is a key biological process that is of increasing importance with the development of a sustainable biofuel industry. The glycoside hydrolase families GH5 (PaMan5A) and GH26 (PaMan26A) endo-β-1,4-mannanases from the coprophilic ascomycete
David Saadoun et al.
Annals of the rheumatic diseases, 73(5), 831-837 (2013-04-23)
The standard-of-care treatment of patients with hepatitis C virus (HCV)-mixed cryoglobulinemia (MC) vasculitis includes pegylated interferon α (PegIFN)-α plus ribavirin and/or rituximab. About 30-40% of patients are non-responders or relapsers to such combination. To analyse the safety and efficacy of
Michael P Manns et al.
Gastroenterology, 147(2), 366-376 (2014-04-15)
MK-5172 is an inhibitor of the hepatitis C virus (HCV) nonstructural protein 3/4A protease; MK-5172 is taken once daily and has a higher potency and barrier to resistance than licensed protease inhibitors. We investigated the efficacy and tolerability of MK-5172
R A Wevers et al.
Clinical chemistry, 40(7 Pt 1), 1245-1250 (1994-07-01)
Although spin-echo techniques are often used to obtain 1H-NMR spectra of serum or plasma samples, they do not provide reliable quantitative analyses of metabolites. We present a standardized procedure, optimized for sensitivity, for using single-pulse 1H-NMR spectroscopy to analyze deproteinized
Ewa Sawosz et al.
International journal of molecular sciences, 16(10), 25214-25233 (2015-10-30)
Our previous studies revealed that graphene had anticancer properties in experiments in vitro with glioblastoma multiforme (GBM) cells and in tumors cultured in vivo. We hypothesized that the addition of arginine or proline to graphene solutions might counteract graphene agglomeration

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service