Skip to Content
Merck
All Photos(3)

Key Documents

W331015

Sigma-Aldrich

Methyl thiobutyrate

greener alternative

natural, 98%, FG

Synonym(s):

S-Methyl thiobutanoate, S-Methyl thiobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2COSCH3
CAS Number:
Molecular Weight:
118.20
FEMA Number:
3310
EC Number:
219-407-1
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.032
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
biological source:
Saccharum officinarum (sugarcane)
food allergen:
no known allergens

biological source

Saccharum officinarum (sugarcane)

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

98%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.461 (lit.)

bp

142-143 °C/757 mmHg (lit.)

density

0.966 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

cheese; musty; sulfurous

SMILES string

CCCC(=O)SC

InChI

1S/C5H10OS/c1-3-4-5(6)7-2/h3-4H2,1-2H3

InChI key

GRLJIIJNZJVMGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Effect of volatile organic compounds from bacteria on nematodes.: Methyl thiobutyrate is identified as a bioactive volatile organic compound in bacterial interactions with nematodes, suggesting its potential use in biocontrol applications in agriculture (Xu et al., 2015).

  • Carboxylesterase activity, cDNA sequence, and gene expression in malathion susceptible and resistant strains of the cotton aphid, Aphis gossypii.: This study explores the enzymatic interactions involving methyl thiobutyrate, providing insights into insecticide resistance mechanisms, which can inform the development of more effective pest management strategies (Pan et al., 2009).


Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service