Skip to Content
Merck
All Photos(1)

Key Documents

W280607

Sigma-Aldrich

Octyl acetate

≥98%, FCC, FG

Synonym(s):

Acetic acid octyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2(CH2)7CH3
CAS Number:
Molecular Weight:
172.26
FEMA Number:
2806
EC Number:
Council of Europe no.:
197
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.007
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

vapor density

5.9 (vs air)

Assay

≥98%

autoignition temp.

515 °F

expl. lim.

8.14 %

refractive index

n20/D 1.418 (lit.)

bp

211 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

floral; fruity; sweet

SMILES string

CCCCCCCCOC(C)=O

InChI

1S/C10H20O2/c1-3-4-5-6-7-8-9-12-10(2)11/h3-9H2,1-2H3

InChI key

YLYBTZIQSIBWLI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Octyl acetate is one of the major constituents of the essential oils of Heracleum crenatifolium, oranges and grapefruit.

Application


  • Update to RIFM fragrance ingredient safety assessment, 3-octyl acetate, CAS registry number 4864-61-3.: This update provides a comprehensive safety assessment of 3-octyl acetate, highlighting its use in fragrances and the ongoing evaluation of its safety for consumer products (Api et al., 2024).

  • Exploring Astrodaucus orientalis (L.) Drude: Phytochemical Analysis and its Biological Potential Against Alzheimer′s and Diabetes.: The study explores the phytochemical composition of Astrodaucus orientalis, including octyl acetate, and its potential therapeutic effects against Alzheimer′s and diabetes, offering new insights into its biochemical applications (Yuca et al., 2024).

  • Energy-efficient recovery of fermented butyric acid using octyl acetate extraction.: This paper presents an energy-efficient method for recovering butyric acid using octyl acetate extraction, showcasing its application in bioprocess engineering and its potential to improve biochemical extraction processes (Oh et al., 2022).

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

189.1 °F - closed cup

Flash Point(C)

87.3 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anticonvulsant activity of furanocoumarins and the essential oil obtained from the fruits of Heracleum crenatifolium.
Tosun F, et al.
Food Chemistry, 107(3), 990-993 (2008)
C Ursin et al.
American Industrial Hygiene Association journal, 56(7), 651-660 (1995-07-01)
A procedure has been developed for measuring the steady state rate of permeation of commercial solvents through living human skin. To get the most consistent results, it was necessary with some solvents to normalize the solvent permeation rate of a
W C Daughtrey et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 12(2), 313-320 (1989-02-01)
The subchronic toxicity of octyl acetate was assessed following its administration to rats via oral gavage, 5 days per week for 13 weeks. Treated rats received undiluted octyl acetate at doses of 0.1, 0.5, or 1.0 g/kg. Control rats received
M J Carroll et al.
The Journal of heredity, 91(1), 68-71 (2000-03-30)
The aliphatic esters octyl acetate and octyl butyrate occur as major components of essential oils in the vittae, or oil tubes, of the wild parsnip (Pastinaca sativa). We determined phenotypic variation and narrow-sense heritabilities of these octyl esters in wild
Citrus flavor. Volatile constituents of the essential oil of the orange (Citrus sinensis).
Bernhard RA.
Journal of Food Science, 26(4), 401-411 (1961)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service