Skip to Content
Merck
All Photos(3)

Key Documents

W274623

Sigma-Aldrich

Dimethyl sulfide

greener alternative

natural, ≥99%, FCC, FG

Synonym(s):

DMS, Methyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2S
CAS Number:
Molecular Weight:
62.13
FEMA Number:
2746
Beilstein:
1696847
EC Number:
Council of Europe no.:
483c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.006
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

300
400

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

vapor density

2.1 (vs air)

vapor pressure

26.24 psi ( 55 °C)
7.79 psi ( 20 °C)

Assay

≥99%

autoignition temp.

402 °F

expl. lim.

19.7 %

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.435 (lit.)

bp

38 °C (lit.)

mp

−98 °C (lit.)

solubility

H2O: soluble 7.28 g/L at 20 °C
ethanol: 95%, clear, colorless to faint yellow or tan (1mL/1mL)

density

0.846 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

green; vegetable; sulfurous; tomato

SMILES string

CSC

InChI

1S/C2H6S/c1-3-2/h1-2H3

InChI key

QMMFVYPAHWMCMS-UHFFFAOYSA-N

Gene Information

General description

Dimethyl sulfide is a volatile flavor compound found in butter, green tea and sweet corn.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-32.8 °F - closed cup

Flash Point(C)

-36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studies on the Flavor of Green Tea.
Kiribuchi, T & Yamanishi T.
Agricultural and Biological Chemistry, 27(1), 56-59 (1963)
Dimethyl sulfide and its precursor in sweet corn.
Bills DD & Keenan TW.
Journal of Agricultural and Food Chemistry, 16(4), 643-645 (1968)
Dimethyl sulfide and the flavor of butter.
Day EA, et al.
Journal of Dairy Science, 47(2), 197-199 (1964)
Sheree Yau et al.
The ISME journal, 7(10), 1944-1961 (2013-04-27)
Organic Lake is a shallow, marine-derived hypersaline lake in the Vestfold Hills, Antarctica that has the highest reported concentration of dimethylsulfide (DMS) in a natural body of water. To determine the composition and functional potential of the microbial community and
Hayley E Arnold et al.
Global change biology, 19(4), 1007-1016 (2013-03-19)
The production of the marine trace gas dimethyl sulfide (DMS) provides 90% of the marine biogenic sulfur in the atmosphere where it affects cloud formation and climate. The effects of increasing anthropogenic CO2 and the resulting warming and ocean acidification

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service