Skip to Content
Merck
All Photos(1)

Key Documents

W243000

Sigma-Aldrich

Ethyl cinnamate

≥98%, stabilized, FCC, FG

Synonym(s):

3-Phenyl-2-propenoic acid ethyl ester, Ethyl 3-phenyl-2-propenoate, NSC 6773

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCOOC2H5
CAS Number:
Molecular Weight:
176.21
FEMA Number:
2430
Beilstein:
775541
EC Number:
Council of Europe no.:
323
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.730
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

contains

synthetic α-tocopherol as stabilizer

refractive index

n20/D 1.558 (lit.)

bp

271 °C (lit.)

mp

6-8 °C (lit.)

density

1.049 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cinnamon; spicy; fruity; balsamic; sweet

SMILES string

CCOC(=O)\C=C\c1ccccc1

InChI

1S/C11H12O2/c1-2-13-11(12)9-8-10-6-4-3-5-7-10/h3-9H,2H2,1H3/b9-8+

InChI key

KBEBGUQPQBELIU-CMDGGOBGSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Decoding the Effect of Running on Flavor Perception Changes during Consumption of Sports Drinks.: This study explores how physical exercise affects the perception of flavors in sports drinks, with ethyl cinnamate playing a role in altering taste perception during and after running (Pu et al., 2024).

  • Ethyl cinnamate suppresses tumor growth through anti-angiogenesis by attenuating VEGFR2 signal pathway in colorectal cancer.: This paper discusses the inhibitory effects of ethyl cinnamate on tumor growth in colorectal cancer by interfering with the VEGFR2 signaling pathway, thereby reducing angiogenesis and tumor progression (Wang et al., 2024).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Astrid Eben et al.
Insects, 11(11) (2020-11-22)
Drosophila suzukii (Diptera: Drosophilidae), is native to southeastern Asia and invaded Europe during the past decade. It causes serious economic damage in cherries and soft fruits. Control strategies rely on few insecticides with varying success. Due to environmental concern, the
Nam-Jin Kim et al.
Pest management science, 64(8), 857-862 (2008-03-08)
This study was aimed at assessing the toxicity of ethyl cinnamate and ethyl p-methoxycinnamate (EMC) identified in Kaempferia galangal L. (Zingiberaceae) rhizome and another 12 known compounds to third-instar larvae from laboratory-reared Culex pipiens pallens Forskal, Aedes aegypti L. and
Rozana Othman et al.
Planta medica, 68(7), 655-657 (2002-07-27)
From the rhizomes of Kaempferia galanga, ethyl cinnamate (EC) was isolated and its vasorelaxant effect was examined on the rat aorta. EC inhibited the tonic contractions induced by high K+ and phenylephrine (PE) in a concentration-dependent manner, with respective IC50
K Bratt et al.
Journal of chemical ecology, 27(11), 2253-2262 (2002-01-31)
Pine weevils (Hylobius abietis) fed less on bark of lodgepole pine (Pinus contorta) than on bark of Scots pine (P. sylvestris). Two pine weevil antifeedants, ethyl trans-cinnamate and ethyl 2,3-dibromo-3-phenyl-propanoate, were isolated from bark of lodgepole pine. These two compounds
Linfang Huang et al.
Journal of ethnopharmacology, 120(1), 123-125 (2008-09-02)
It is well known that fragrance impacts behaviors and autonomic functions, and is increasingly used as relaxant, carminative, as well as sedative in aromatherapy. Kaempferia galanga L. is one of the popular traditional aromatic medicinal plants used in tropics and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service