Skip to Content
Merck
All Photos(1)

Documents

W236403

Sigma-Aldrich

Decanoic acid

≥99.5%, FCC, FG

Synonym(s):

1-Nonanecarboxylic acid, Acid C10, C10:0, Capric acid, NSC 5025

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8COOH
CAS Number:
Molecular Weight:
172.26
FEMA Number:
2364
Beilstein:
1754556
EC Number:
Council of Europe no.:
11
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.011
NACRES:
NA.21

biological source

palm oil

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.210

vapor pressure

15 mmHg ( 160 °C)

Assay

≥99.5%

bp

268-270 °C (lit.)

mp

27-32 °C (lit.)

density

0.893 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fatty; citrus; rancid; sour

SMILES string

CCCCCCCCCC(O)=O

InChI

1S/C10H20O2/c1-2-3-4-5-6-7-8-9-10(11)12/h2-9H2,1H3,(H,11,12)

InChI key

GHVNFZFCNZKVNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Decanoic acid, a medium-chain fatty acid, is one of the volatile flavor/aroma constituents that has been identified in apple wine, stored nonfat dry milk and parmesan cheese.

Application


  • Chemical Characterization of Terpene-Based Hydrophobic Eutectic Solvents and Their Application for Pb(II) Complexation during Solvent Extraction Procedure.: This paper discusses the use of Decanoic acid in novel eutectic solvents for heavy metal extraction, presenting a green chemistry approach to environmental cleanup and recovery of valuable metals (Suljkanović et al., 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

296.6 °F - closed cup

Flash Point(C)

147 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid Analysis of Flavor Volatiles in Apple Wine Using Headspace Solid?Phase Microextraction
Wang L, et al.
Journal of the Institute of Brewing, 110(1), 57-65 (2004)
Optimization of solid phase microextraction analysis for the headspace volatile compounds of Parmesan cheese
Lee JH, et al.
Journal of Agricultural and Food Chemistry, 51(5), 1136-1140 (2003)
Yonca Karagül-Yüceer et al.
Journal of agricultural and food chemistry, 50(2), 305-312 (2002-01-10)
Nonfat dry milk (NDM) is widely used both as an ingredient in other preparations and for direct consumption. Flavor quality of NDM is a critical parameter because it can directly impact final product quality. Flavors can be formed in NDM
Morteza Moradi et al.
Journal of chromatography. A, 1229, 30-37 (2012-02-07)
A new, efficient and environmentally friendly method for the analysis of parabens as model compounds was developed using solidified floating vesicular coacervative drop microextraction (SFVCDME). A supramolecular solvent consisting of vesicles of decanoic acid in the nano- and microscale regimes
Hiroyuki Kaiya et al.
Endocrinology, 143(9), 3454-3463 (2002-08-24)
In this study, we report the purification, cDNA cloning, and characterization of the novel growth hormone-releasing peptide, ghrelin, in the chicken (Gallus gallus). Chicken ghrelin is composed of 26 amino acids (GSSFLSPTYKNIQQQKDTRKPTARLH) and possesses 54% sequence identity with human ghrelin.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service