Key Documents
TEM-HC200-NI-100
Sigma-Aldrich
Holey Carbon Film Supported Nickel Grids
size 200 mesh, hole size 100 μm, box of 25
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Pricing and availability is not currently available.
packaging
box of 25
hole size
100 μm
size
200 mesh
Compare Similar Items
View Full Comparison
Show Differences
1 of 4
This Item
TEM-HC300-NI-100TEM-HC200-AU-100TEM-HC400-NI-100
hole size
100 μm
hole size
100 μm
hole size
100 μm
hole size
100 μm
size
200 mesh
size
300 mesh
size
200 mesh
size
400 mesh
General description
Holey Carbon Film Supported Nickel Grids, 200 Mesh, Average hole size is 100 micron, 25/box
Choose from one of the most recent versions:
Certificates of Analysis (COA)