Skip to Content
Merck
All Photos(4)

Key Documents

T72761

Sigma-Aldrich

Trimethylamine hydrochloride

98%

Synonym(s):

Trimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N · HCl
CAS Number:
Molecular Weight:
95.57
Beilstein:
3905063
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

283-284 °C (dec.) (lit.)

SMILES string

Cl[H].CN(C)C

InChI

1S/C3H9N.ClH/c1-4(2)3;/h1-3H3;1H

InChI key

SZYJELPVAFJOGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Trimethylamine hydrochloride is a precursor to prepare aluminium chloride- trimethylamine hydrochloride ionic liquid, which is commonly used in electrodeposition of aluminum wires. It is also used in the synthesis of trimethylamine gallane, trimethylamine-borane and azido-bridged perovskite-type metal-organic frameworks (MOFs).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

GaN@ ZIF-8: Selective formation of gallium nitride quantum dots inside a zinc methylimidazolate framework.
Esken D, et al.
Journal of the American Chemical Society, 133(41), 16370-16373 (2011)
Electrodeposition of aluminum wires from the Lewis acidic AlCl3/trimethylamine hydrochloride ionic liquid without using a template.
Su C-J, et al.
Electrochemical Communications, 34, 170-173 (2013)
Danny Orabi et al.
JCI insight, 6(9) (2021-05-15)
Gut microbe-derived metabolites influence human physiology and disease. However, establishing mechanistic links between gut microbial metabolites and disease pathogenesis in animal models remains challenging. The major route of absorption for microbe-derived small molecules is venous drainage via the portal vein
Electrodeposition of purified aluminum coatings from dimethylsulfone-AlCl3 electrolytes with trimethylamine hydrochloride.
Miyake M, et al.
Surface and Coatings Technology, 206(19-20), 4225-4229 (2012)
Cation-dependent magnetic ordering and room-temperature bistability in azido-bridged perovskite-type compounds.
Zhao X-H, et al.
Journal of the American Chemical Society, 135(43), 16006-16009 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service