Skip to Content
Merck
All Photos(2)

Documents

T49484

Sigma-Aldrich

Tri-n-butylphosphine

97%

Synonym(s):

Tributylphosphine, NSC 91700, P(n-Bu)3, TBP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]3P
CAS Number:
Molecular Weight:
202.32
Beilstein:
1738261
EC Number:
MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

vapor density

9 (vs air)

Quality Level

Assay

97%

autoignition temp.

392 °F

reaction suitability

reagent type: ligand
reaction type: Acetylations

reagent type: ligand
reaction type: Addition Reactions

reagent type: ligand
reaction type: Stille Coupling

refractive index

n20/D 1.462 (lit.)

bp

150 °C/50 mmHg (lit.)

density

0.81 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

CCCCP(CCCC)CCCC

InChI

1S/C12H27P/c1-4-7-10-13(11-8-5-2)12-9-6-3/h4-12H2,1-3H3

InChI key

TUQOTMZNTHZOKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

1,4-addition catalyst; used with disulfides for the thioetherification of alcohols; acylation catalyst; used to prepare active esters.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1A

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

242.6 °F - closed cup

Flash Point(C)

117 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing-Yu Wu et al.
The Journal of organic chemistry, 73(22), 9137-9139 (2008-10-24)
Aziridines underwent cyclization reaction with carbon disulfide and isothiocyanate in the presence of organophosphine to afford thiazolidinone derivatives in good to high yields. The mechanistic study revealed that organophosphine serves as a catalyst in the reaction.
J X Yan et al.
Journal of chromatography. A, 813(1), 187-200 (1998-08-11)
A simple technique is introduced to identify and quantitate cysteine (Cys) after acid hydrolysis of protein. The technique involves using 9-fluorenylmethyl chloroformate (Fmoc)-based amino acid analysis that recovers all of the amino acids (asparagine and glutamine are recovered in their
E Kaniowska et al.
Journal of chromatography. A, 798(1-2), 27-35 (1998-05-02)
Several human diseases, in particular metabolic disorders, often lead to the accumulation of characteristic metabolites in plasma, urine and cells. The selected diseases of this type include cystinuria and homocystinuria. In the typical laboratory diagnosis of these two diseases, a
Yin-Wei Sun et al.
Organic letters, 12(24), 5664-5667 (2010-11-13)
P(n)Bu(3)-catalyzed cyclization reactions of salicylaldimines and salicylaldehydes with ethyl 2,3-butadienoate gave the corresponding functionalized chromans in moderate to good yields in THF under mild conditions. The new reaction provides a new method for the synthesis of biologically active chroman products.
Kersti B Nilsson et al.
Dalton transactions (Cambridge, England : 2003), (9)(9), 1312-1319 (2004-07-15)
The coordination chemistry of the solvate complexes of the relatively soft electron-pair acceptor copper(I) has been studied in solution and solid state in seven solvents with strong electron-pair donor properties, liquid ammonia, trimethyl, triethyl, triisopropyl, tri-n-butyl and triphenyl phosphite, and

Articles

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service