Skip to Content
Merck
All Photos(1)

Key Documents

T32603

Sigma-Aldrich

2-Thiophenecarboxylic acid

ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4O2S
CAS Number:
Molecular Weight:
128.15
Beilstein:
110150
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

product line

ReagentPlus®

Assay

99%

bp

260 °C (lit.)

mp

125-127 °C (lit.)

SMILES string

OC(=O)c1cccs1

InChI

1S/C5H4O2S/c6-5(7)4-2-1-3-8-4/h1-3H,(H,6,7)

InChI key

QERYCTSHXKAMIS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Matthew F McCown et al.
Antimicrobial agents and chemotherapy, 53(5), 2129-2132 (2009-03-11)
In vitro, telaprevir selects subtype-specific resistance pathways for hepatitis C virus GT-1a and GT-1b, as described to have occurred in patients. In GT-1a, the HCV-796 resistance mutation C316Y has low replication capacity (7%) that can be compensated for by the
Hao Li et al.
Organic letters, 13(14), 3682-3685 (2011-06-17)
A stereocontrolled synthesis of α-amino-α'-alkoxy ketones is described. This pH-neutral copper(I) thiophene-2-carboxylate (CuTC)-catalyzed cross-coupling of amino acid thiol esters and chiral nonracemic α-alkoxyalkylstannanes gives α-amino-α'-alkoxy ketones in good to excellent yields with complete retention of configuration at the α-amino- and
C Savarin et al.
Organic letters, 3(14), 2149-2152 (2001-07-07)
[reaction: see text] A new protocol for the palladium-catalyzed, copper-mediated coupling of aryl and alkenyl iodides with boronic acids is described. As an alternative to the well-known and widely used Suzuki cross-coupling, this reaction occurs in the absence of a
Kerim Babaoglu et al.
Nature chemical biology, 2(12), 720-723 (2006-10-31)
Fragment-based screens test multiple low-molecular weight molecules for binding to a target. Fragments often bind with low affinities but typically have better ligand efficiencies (DeltaG(bind)/heavy atom count) than traditional screening hits. This efficiency, combined with accompanying atomic-resolution structures, has made
Laval Chan et al.
Bioorganic & medicinal chemistry letters, 14(3), 797-800 (2004-01-27)
Further SAR studies on the thiophene-2-carboxylic acids are reported. These studies led to the identification of a series of tertiary amides that show inhibition of both HCV NS5B polymerase in vitro and HCV subgenomic RNA replication in Huh-7 cells. Structural

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service