SYNF301005
Synthware™ single neck round bottom flask
capacity 5 mL, joint: ST/NS 14/10
Synonym(s):
Synthware F301005, glass round flask
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
Recommended Products
material
borosilicate glass flask
round bottom
single-neck
Joint
joint: ST/NS14/10
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F301005
capacity
5 mL
Related Categories
General description
Supplied with one holed compression caps, one Viton® O-rings and one PTFE - faced silicone rubber septum. For replacement caps, see RC112; for Viton® O-rings, see R014; for septa, see R1420.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)