Skip to Content
Merck

SYNC625300

Synthware Allihn condenser

joint: ST/NS 29/42, jacket L 300 mm, hose size 10 mm

Synonym(s):

Synthware C625300, straight condenser

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.51
UNSPSC Code:
41121800
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


joint

joint: ST/NS29/42

packaging

pkg of 1 ea

manufacturer/tradename

Synthware C625300

hose size

10 mm

jacket L

300 mm

General description

“Synthware” Allihn condenser features a 300 mm jacketed length for superior cooling efficiency and removable hose connections. A series of internal glass bulbs increase surface area for efficient cooling and prevent loss of volatile substances. With upper standard taper outer joint (29/42) to fit various laboratory glassware and a lower drip tip joint.

Legal Information

Synthware is a trademark of Kemtech America Inc.


Still not finding the right product?

Explore all of our products under Synthware Allihn condenser




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library