Merck
SYNC182403
Synthware chromatography reservoir
capacity 500 mL, joint: ST/NS 24/40
Synonym(s):
Synthware C182403, Glass chromatography reservoir
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.51
UNSPSC Code:
41121800
Manufacturer/tradename:
Synthware C182403
Capacity:
500 mL
Material:
colorless , glass
Sterility:
non-sterile
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
colorless , glass
joint
joint: ST/NS24/40
sterility
non-sterile
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C182403
capacity
500 mL
suitability
suitable for chromatography
General description
For flash chromatography.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)