Merck
SYNC159190
Synthware West condenser
joint: ST/NS 19/22, jacket L 190 mm
Synonym(s):
Synthware C159190, glass West condenser
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.51
UNSPSC Code:
41121800
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C159190
jacket L
190 mm
General description
With narrow annular space for fast coolant speed, thus high cooling efficiency.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Still not finding the right product?
Explore all of our products under Synthware West condenser
Choose from one of the most recent versions:
Certificates of Analysis (COA)