Merck
Skip To
SYNC1324
Synthware chromatography flow control adapter
joint: ST/NS 24/40, with PTFE stopcock
Synonym(s):
Synthware C1324, Glass chromatography adapter
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
About This Item
NACRES:
NB.51
UNSPSC Code:
41121800
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C1324
suitability
suitable for chromatography
General description
0-4 mm high vacuum valve with metering tip to adjust air pressure. With standard taper joint. Order CC3525 pinch type clamps with screw-locking device for ball joints. For replacement O-rings on high vacuum plug shaft, order R010. The O-rings on the high vacuum PTFE valve are completely encapsulated with thin layer of PTFE, making it inert to chemicals.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
SYNA392440SYNC1301SYNA391420
joint
joint: ST/NS24/40
joint
joint: ST/NS24/40
joint
-
joint
joint: ST/NS14/20
manufacturer/tradename
Synthware C1324
manufacturer/tradename
Synthware A392440
manufacturer/tradename
Synthware C1301
manufacturer/tradename
Synthware A391420
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
suitability
suitable for chromatography
suitability
-
suitability
suitable for chromatography
suitability
-
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)