Skip to Content
Merck
All Photos(4)

Documents

P56607

Sigma-Aldrich

Pyrazole

98%

Synonym(s):

1,2-Diazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4N2
CAS Number:
Molecular Weight:
68.08
Beilstein:
103775
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

crystals

bp

186-188 °C (lit.)

mp

67-70 °C (lit.)

SMILES string

c1cn[nH]c1

InChI

1S/C3H4N2/c1-2-4-5-3-1/h1-3H,(H,4,5)

InChI key

WTKZEGDFNFYCGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used as a ligand to prepare organometallic compounds.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 1

Target Organs

spleen,Thyroid

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transition Met. Chem. (London), 19, 165-165 (1994)
Heterocycles, 37, 2087-2087 (1994)
Roberta Zaninetti et al.
ChemMedChem, 8(4), 633-643 (2013-02-26)
Combretastatin A1 (CA1) binds to the β-subunit at the colchicine binding site of tubulin and inhibits polymerization. As such, it is both an antitumor agent and a vascular disrupting agent. It has been shown to be at least tenfold more
Mohamed Jawed Ahsan et al.
Bioorganic & medicinal chemistry letters, 22(23), 7029-7035 (2012-10-27)
A series of 43, 3a,4-dihydro-3H-indeno[1,2-c]pyrazole-2-carboxamide/carbothioamide analogues (D01-D43) were analysed using Petra, Osiris, Molinspiration and ALOGPS (POMA) to identify pharmacophore, toxicity prediction, lipophilicity and bioactivity. All the compounds were evaluated for anti-HIV activity. 3-(4-Chlorophenyl)-N-(4-fluorophenyl)-6,7-dimethoxy-3a,4-dihydro-3H-indeno[1,2-c]pyrazole-2-carboxamide (D07) was found to be the most
Cinzia Chiappe et al.
The journal of physical chemistry. B, 117(2), 668-676 (2012-12-21)
Ionic liquids (ILs) composed of two different pyrazolium cations with dicyanamide and bis(trifluoromethanesulfonyl)imide anions have been synthesized and characterized by NMR, Kamlet-Taft solvatochromic parameters, conductivity and rheological measurements, as well as ab initio calculations. Density functional calculations for the two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service