Skip to Content
Merck
All Photos(1)

Key Documents

P31205

Sigma-Aldrich

Phenylpropiolic acid

99%

Synonym(s):

Phenylpropynoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C≡CCOOH
CAS Number:
Molecular Weight:
146.14
Beilstein:
742587
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

crystals

mp

135-137 °C (lit.)

storage temp.

2-8°C

SMILES string

OC(=O)C#Cc1ccccc1

InChI

1S/C9H6O2/c10-9(11)7-6-8-4-2-1-3-5-8/h1-5H,(H,10,11)

InChI key

XNERWVPQCYSMLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phenylpropiolic acid can:
  • React with 2-tert-butoxypyridine in the presence of boron trifluoride·diethyl etherate to form the corresponding tert-butyl ester.
  • Undergo decarboxylative coupling with aryl halides such as p-chloroiodobenzene and 1-chloro-4-iodobenzene.
  • Undergo halodecarboxylation to form 1-haloalkynes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth Joubert et al.
Food chemistry, 136(2), 1078-1085 (2012-11-06)
Z-2-(β-d-glucopyranosyloxy)-3-phenylpropenoic acid (PPAG), a compound postulated to contribute to the taste and mouthfeel of fermented rooibos tea (Aspalathus linearis), was isolated from unfermented rooibos plant material. Its structure was unequivocally confirmed by LC-MS, -MS(2), FT-IR and NMR of the underivatised
A fast and practical synthesis of tert-butyl esters from 2-tert-butoxypyridine using boron trifluoride? diethyl etherate under mild conditions.
La M T and Kim H K
Tetrahedron, 74(27), 3748-3754 (2018)
Kun Li et al.
Ying yong sheng tai xue bao = The journal of applied ecology, 21(7), 1779-1784 (2010-10-01)
Taking the tissue-cultured seedlings of grape cultivar Red Globe as test objects, this paper examined the effects of their root aqueous extracts on seedling's growth, with the allelochemicals identified by LC-MS. The results showed that 0.02 g x ml(-1) (air-dried
Y Q Feng et al.
Se pu = Chinese journal of chromatography, 18(3), 224-228 (2003-01-25)
Glycine derivatized beta-cyclodextrin bonded silica (GCDS) has been prepared for high performance liquid chromatography through the reactions of beta-cyclodextrin bonded silica with tosyl chloride and glycine in sequence. The separation performance of GCDS for positional isomers, dansyl amino acids and
Iwona Morkunas et al.
Journal of plant physiology, 168(5), 424-433 (2010-11-09)
This study investigated the effects of cross-talk interactions of sucrose and infection caused by a pathogenic fungus Fusarium oxysporum f.sp. lupini on the regulation of the phenylpropanoid pathway, i.e. the level of expression of genes encoding enzymes participating in flavonoid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service