P2000
Pentachloroethane
95%
Synonym(s):
1,1,1,2,2-Pentachloroethane, HCC 120, Pentalin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CCl3CHCl2
CAS Number:
Molecular Weight:
202.29
Beilstein:
1736845
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
refractive index
n20/D 1.502 (lit.)
bp
161-162 °C (lit.)
density
1.68 g/mL at 25 °C (lit.)
SMILES string
ClC(Cl)C(Cl)(Cl)Cl
InChI
1S/C2HCl5/c3-1(4)2(5,6)7/h1H
InChI key
BNIXVQGCZULYKV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - STOT RE 1
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jens Christian Nielsen et al.
Applied microbiology and biotechnology, 101(22), 8237-8248 (2017-10-11)
The Brazilian sugarcane industry constitutes one of the biggest and most efficient ethanol production processes in the world. Brazilian ethanol production utilizes a unique process, which includes cell recycling, acid wash, and non-aseptic conditions. Process characteristics, such as extensive CO2
Alex M Zimmer et al.
The Journal of experimental biology, 223(Pt 2) (2019-12-20)
Freshwater fishes absorb Na+ from their dilute environment using ion-transporting cells. In larval zebrafish (Danio rerio), Na+ uptake is coordinated by (1) Na+/H+ exchanger 3b (Nhe3b) and (2) H+-ATPase-powered electrogenic uptake in H+-ATPase-rich (HR) cells and by (3) Na+-Cl--cotransporter (Ncc)
Veny Uppal et al.
Journal of food science and technology, 49(2), 184-191 (2013-04-11)
Germination of legumes followed by hydrothermal treatments is an effective mean of improving nutritive value of legumes. The protein content of mungbean, chickpea and cowpea increased by 9-11, 11-16 and 8-11% after germination. A significant (p ≤ 0.05) decrease in protein content
A M Allen et al.
Gut, 33(3), 392-396 (1992-03-01)
Endogenous and exogenous stimulation of the pancreas was studied to determine whether changes in protein output could be linked to decreased total plasma amino acid concentrations. In fasted rats, diversion of pancreatic juice resulted in a transient increase in protein
P Melin et al.
Journal of applied microbiology, 100(2), 264-271 (2006-01-25)
Investigate the survival of liquid formulations of the biocontrol yeast Pichia anomala J121 at different temperatures, and develop a system for comparative studies of different storage conditions and formulations. The survival of P. anomala in liquid formulations with lactose, starch
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service