Skip to Content
Merck
All Photos(1)

Key Documents

O4733

Sigma-Aldrich

Octanoyl chloride

99%

Synonym(s):

Capryloyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6COCl
CAS Number:
Molecular Weight:
162.66
Beilstein:
635917
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

2.5 mmHg ( 20 °C)

Assay

99%

refractive index

n20/D 1.435 (lit.)

bp

195 °C (lit.)

density

0.953 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCC(Cl)=O

InChI

1S/C8H15ClO/c1-2-3-4-5-6-7-8(9)10/h2-7H2,1H3

InChI key

REEZZSHJLXOIHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Applications of octanoyl chloride include:
  • Synthesis of (R)-2-propyloctanoic acid, a therapeutic agent for Alzheimer′s disease.
  • Synthesis of a variety of N-n-octyl-D-gluconamide based organogels.
  • Building 2-heptylbenzo[d]thiazole moiety to synthesize conjugated copolymer with fluorine, as cathode interlayer in inverted polymer solar cells (PSCs).
  • Octanoyl chloride is one of the essential precursors in the total synthesis of (−)-mandelalide L, a marine macrolide that display significant cytotoxicity against human cancer cell lines.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic Access to the Mandelalide Family of Macrolides: Development of an Anion Relay Chemistry Strategy.
Nguyen M H, et al.
The Journal of Organic Chemistry, 83(8), 4287-4306 (2018)
Organogels from carbohydrate amphiphiles.
Hafkamp R J, et al.
The Journal of Organic Chemistry, 64(2), 412-426 (1999)
5, 6-Difluorobenzothiazole-Based Conjugated Polymers with Large Band Gaps and Deep Highest Occupied Molecular Orbital Levels.
Sun J, et al.
ACS Applied Materials & Interfaces, 10(13), 11094-11100 (2018)
Shaopeng Wu et al.
Materials science & engineering. C, Materials for biological applications, 71, 973-981 (2016-12-19)
To understand the effects of GRGDF modification on MC3T3-E1 cell behavior, we cultured these cells onto a biomimetic liquid crystalline matrix modified with GRGDF peptide (OPC-GA-RGD). Successful immobilization of GRGDF on the liquid crystalline surface was verified by fluorescent labeling
M Thirumalai et al.
Journal of chromatography. A, 1569, 62-69 (2018-07-22)
In this article, a new reversed-phase high-performance liquid chromatography (RP-HPLC) method for the rapid, selective and sequential separation of toxic heavy metal ions namely, Pb2+, Hg2+, and Cd2+ is reported. For this RP-HPLC method, a C18 silica monolith column (Chromolith

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service