Skip to Content
Merck
All Photos(1)

Key Documents

N22908

Sigma-Aldrich

3-Nitropropionic acid

97%

Synonym(s):

β-Nitropropionic acid, 3-Nitropropanoic acid, Bovinocidin, Hiptagenic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2N(CH2)2CO2H
CAS Number:
Molecular Weight:
119.08
Beilstein:
1759889
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

crystals

mp

68-70 °C (lit.)

storage temp.

2-8°C

SMILES string

OC(CC[N+]([O-])=O)=O

InChI

1S/C3H5NO4/c5-3(6)1-2-4(7)8/h1-2H2,(H,5,6)

InChI key

WBLZUCOIBUDNBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Excitotoxin shown to cause brain lesions similar to those of Huntington′s disease.

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emmanuel Brouillet et al.
Journal of neurochemistry, 95(6), 1521-1540 (2005-11-23)
Huntington's disease (HD) is a neurodegenerative disorder caused by a mutation in the gene encoding Huntingtin. The mechanisms underlying the preferential degeneration of the striatum, the most striking neuropathological change in HD, are unknown. Of those probably involved, mitochondrial defects
Samantha Corrà et al.
iScience, 26(10), 107955-107955 (2023-10-09)
Mutations in MPV17 are a major contributor to mitochondrial DNA (mtDNA) depletion syndromes, a group of inherited genetic conditions due to mtDNA instability. To investigate the role of MPV17 in mtDNA maintenance, we generated and characterized a Drosophila melanogaster Mpv17
E Brouillet et al.
Current opinion in neurology, 8(6), 469-473 (1995-12-01)
Compelling evidence suggests that a defect in energy metabolism may play a role in the pathogenesis of various degenerative disorders including Parkinson's disease and Huntington's disease. The behavioural and neuropathological consequences in primates of chronic systemic administration of mitochondrial toxins
Rajat Sandhir et al.
Biochimica et biophysica acta, 1832(3), 421-430 (2012-12-12)
The study was designed to investigate the beneficial effect of quercetin supplementation in 3-nitropropionic acid (3-NP) induced model of Huntington's disease (HD). HD was induced in rats by administering sub-chronic dose of 3-NP, intraperitoneally, twice daily for 17days. Quercetin was
Wang-Tso Lee et al.
Progress in neurobiology, 72(2), 87-110 (2004-04-06)
Huntington's disease (HD) is an inherited neurodegenerative disease, in which there is progressive motor and cognitive deterioration, and for which the pathogenesis of neuronal death remains controversial. Mitochondrial toxins like 3-nitropropionic acid (3-NP) and malonate, functioning as the inhibitors of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service