All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C15H16O4S2
CAS Number:
Molecular Weight:
324.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
form
solid
SMILES string
[S](=O)(=O)(C[S](=O)(=O)c2ccc(cc2)C)c1ccc(cc1)C
InChI
1S/C15H16O4S2/c1-12-3-7-14(8-4-12)20(16,17)11-21(18,19)15-9-5-13(2)6-10-15/h3-10H,11H2,1-2H3
InChI key
XLOXYWLRAKCDQL-UHFFFAOYSA-N
Other Notes
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Sandi A Kwee et al.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging, 19(3), 446-455 (2016-10-28)
[ Tumor and adjacent liver were profiled by liquid chromatography mass spectrometry, quantifying phosphatidylcholine species by mass-to-charge ratio. For clinical-radiopathologic correlation, HCC profiles were reduced to two orthogonal principal component factors (PCF1 and PCF2) accounting for 80 % of total profile
Sandi A Kwee et al.
Annals of nuclear medicine, 26(6), 501-507 (2012-05-03)
To evaluate fluorine-18 fluorocholine (FCH) PET/CT for the detection of recurrent prostate cancer in relation to prostate-specific antigen (PSA) level. FCH PET/CT was performed in 50 patients with rising PSA levels at follow-up of primary treatment of prostate cancer (radical
Sandi Kwee et al.
Clinical and translational science, 5(1), 65-70 (2012-03-02)
To examine the effects of chemotherapy on circulating cell-free DNA (cfDNA) composition in relation to investigational whole-body measurement of tumor activity by fluorine-18 fluorocholine (FCH) positron emission tomography/computed tomography (PET/CT) in hormone-refractory prostate cancer (HRPC). Serial FCH PET/CT scans were
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service