Skip to Content
Merck
All Photos(1)

Documents

M84402

Sigma-Aldrich

3-Methylthiophene

98%

Synonym(s):

β-Methylthiophene, 3-Thienylmethane, 3-Thiotolene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6S
CAS Number:
Molecular Weight:
98.17
Beilstein:
1300
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

42.4 mmHg ( 37.7 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.519 (lit.)

bp

114 °C/738 mmHg (lit.)

mp

−69 °C (lit.)

density

1.016 g/mL at 25 °C (lit.)

SMILES string

Cc1ccsc1

InChI

1S/C5H6S/c1-5-2-3-6-4-5/h2-4H,1H3

InChI key

QENGPZGAWFQWCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Conducting polymer precursor.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Linlin Li et al.
Materials science & engineering. C, Materials for biological applications, 53, 36-42 (2015-06-05)
A polymer film incorporated gold nanoparticle modified electrode was fabricated. The fabricated process involved eletrodeposition of gold nanoparticles and electropolymerization of the 3-methylthiophene (abbreviated 3MT) onto the glassy carbon electrode (GCE). The resulting electrode (P3MT-nano-Au/GCE) was characterized by scanning electron
Journal of the American Chemical Society, 117, 233-233 (1995)
Nannan Jian et al.
Physical chemistry chemical physics : PCCP, 21(13), 7174-7182 (2019-03-20)
Conjugated fluorophores have been extensively used for fluorescence sensing of various substances in the field of life processes and environmental science, due to their noninvasiveness, sensitivity, simplicity and rapidity. Most existing conjugated fluorophores exhibit excellent light-emitting performance in dilute solutions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service