Skip to Content
Merck
All Photos(1)

Documents

L5632

Sigma-Aldrich

Lupeol

≥94%

Synonym(s):

β-Viscol, 20(29)-Lupen-3β-ol, 3β-Hydroxy-20(29)-lupene, Clerodol, Fagarasterol, Lupenol, Monogynol B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H50O
CAS Number:
Molecular Weight:
426.72
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

≥94% (TLC)
≥94%

storage temp.

2-8°C

SMILES string

[H]C12CCC3C4(C)CCC(O)C(C)(C)[C@]4([H])CCC3(C)[C@]1(C)CCC5(C)CC[C@H](C(C)=C)[C@]25[H]

InChI

1S/C30H50O/c1-19(2)20-11-14-27(5)17-18-29(7)21(25(20)27)9-10-23-28(6)15-13-24(31)26(3,4)22(28)12-16-30(23,29)8/h20-25,31H,1,9-18H2,2-8H3/t20-,21+,22-,23+,24-,25+,27+,28-,29+,30+/m0/s1

InChI key

MQYXUWHLBZFQQO-QGTGJCAVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lupeol is a novel anti-cancer and anti-inflammatory dietary triterpene. They help in stabilizing phospholipid bilayers in plant cell membranes. In general, this group contains 28-29 carbons along with carbon-carbon double bonds, usually one in the sterol nucleus and at times a second in the alkyl side chain.

Application

Lupeol was used as reference standard in method development and validation of lupeol from Vernonia Cinerea using HPLC method. It has been used as a standard for the identification and quantification of triterpenoids in blueberry cuticular wax by GC.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuang Liang et al.
Journal of natural products, 73(4), 532-535 (2010-03-03)
Phytochemical examination of the methanolic extract from the stem bark of Daphne aurantiaca led to the isolation of six new sesquiterpenoids, dauca-3,11-dien-2alpha,15-diol (1), 3-oxoguai-4-ene-11,12-diol (2), 4alpha,5alpha,8alpha,11alphaH-3-oxoguai-1(10)-en-12,8-olide-7alpha-diol (3), 4alpha,5alpha,8alpha,11betaH-3-oxoguai-1(10)-en-12,8-olide-7beta-diol (4), 4alpha,5betaH-guai-9,7(11)-dien-12,8-olide-1alpha,8alpha-diol (5), 4alpha,5alphaH-guai-9,7(11)-dien-12,8-olide-1alpha,8alpha-diol (6), and a new diterpenoid, 12-O-benzoylphorbol 13-nonanoate (7)
Fruit characteristics and cuticle triterpenes as related to postharvest quality of highbush blueberries.
Moggia C, et al.
Sci. Hortic., 211, 449-457 (2016)
Anupam Maurya et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 47(1), 152-161 (2012-06-05)
Two triterpenoids ursolic acid (1) and lupeol (2) isolated and characterized from Eucalyptus tereticornis and Gentiana kurroo were subjected to in silico QSAR modeling and docking studies and later the predicted results were confirmed through in vivo experiments. QSAR modeling
Hifzur Rahman Siddique et al.
Life sciences, 88(7-8), 285-293 (2010-12-02)
Since ancient times, natural products have been used as remedies to treat human diseases. Lupeol, a phytosterol and triterpene, is widely found in edible fruits, and vegetables. Extensive research over the last three decades has revealed several important pharmacological activities
Development and validation of High Performance Liquid Chromatographic Method for the Simultaneous Determination of ?-sitosterol and Lupeol in Veronica cinerea Linn.
Shah, Willy, M. B. Kekare, and Vikas Vaidya.
International Journal of Pharma and Bio Sciences, 1 (3), 1-5 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service