Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
[CH3CH(OH)COO]2Ca · xH2O
CAS Number:
Molecular Weight:
218.22 (anhydrous basis)
UNSPSC Code:
12352207
NACRES:
NA.22
PubChem Substance ID:
EC Number:
248-953-3
Beilstein/REAXYS Number:
4923285
MDL number:
Assay:
~90%
assay
~90%
Quality Segment
storage temp.
2-8°C
SMILES string
O.C[C@H](O)C(=O)O[Ca]OC(=O)[C@H](C)O
InChI
1S/2C3H6O3.Ca.H2O/c2*1-2(4)3(5)6;;/h2*2,4H,1H3,(H,5,6);;1H2/q;;+2;/p-2/t2*2-;;/m00../s1
InChI key
MDAVASCOAJMZHZ-NVKWYWNSSA-L
Application
Calcium L-lactate hydrate has been used as a sole carbon and energy source, to study the sulfate reduction ability of Desulfovibrio desulfuricans under different conditions.
Still not finding the right product?
Explore all of our products under Calcium L-lactate hydrate
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
