Skip to Content
Merck
All Photos(1)

Documents

I1804

Sigma-Aldrich

Indan

95%

Synonym(s):

Hydrindene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10
CAS Number:
Molecular Weight:
118.18
Beilstein:
1904376
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.537 (lit.)

bp

176 °C (lit.)

mp

−51 °C (lit.)

density

0.965 g/mL at 25 °C (lit.)

SMILES string

C1Cc2ccccc2C1

InChI

1S/C9H10/c1-2-5-9-7-3-6-8(9)4-1/h1-2,4-5H,3,6-7H2

InChI key

PQNFLJBBNBOBRQ-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Li et al.
Chemical communications (Cambridge, England), 46(22), 3979-3981 (2010-04-22)
(S,R)-Indan-ambox ligand and its ruthenium(II) complex have been prepared and successfully applied to asymmetric hydrogenation of prochiral simple ketones. A wide range of unfunctionalized ketones are reduced by Ru(II)-indan-ambox catalyst with excellent enantioselectivities (up to 97% ee).
Kenji Kabashima et al.
FEBS letters, 578(1-2), 36-40 (2004-12-08)
Nuclear factor kappa B (NF-kappaB) plays a wide variety of pathophysiological roles and modulation of its pathway can be a good novel drug target. Here, we found that our recently synthesized NF-kappaB inhibitor attenuated an ovalbumin-specific delayed-type hypersensitivity response in
Peter R Andreana et al.
Organic letters, 6(23), 4231-4233 (2004-11-05)
A catalytic asymmetric Passerini reaction using tridentate indan (pybox) Cu(II) Lewis acid complex 4 with substrates capable of bidentate coordination has been achieved. The reaction occurs via ligand-accelerated catalysis.
Mónica López-García et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 10(12), 3006-3014 (2004-06-24)
The efficient chemoenzymatic synthesis of enantiopure 1,3-difunctionalized indane derivatives has been achieved. The corresponding cis and trans N-protected amino alcohols were successfully resolved by acetylation using lipase B, which is a biocatalyst isolated from Candida antarctica. All the possible isomers
Sarathy Kesavan et al.
Organic letters, 9(25), 5203-5206 (2007-11-14)
Alkylidene indane and ring-expanded scaffolds have been prepared using an enantioselective crotylation/Heck cyclization sequence. Further diversification using consecutive cyclopropanation-Cope rearrangement affords novel chemotypes including spiroindane frameworks.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service