Skip to Content
Merck
All Photos(2)

Key Documents

I12409

Sigma-Aldrich

α-Ionone

technical grade, 90%

Synonym(s):

alpha-Ionone, 4-(2,6,6-Trimethyl-2-cyclohexenyl)-3-buten-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30
Beilstein:
3197885
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

liquid

refractive index

n20/D 1.498 (lit.)

bp

259-263 °C (lit.)

density

0.93 g/mL at 25 °C (lit.)

SMILES string

CC(=O)\C=C\C1C(C)=CCCC1(C)C

InChI

1S/C13H20O/c1-10-6-5-9-13(3,4)12(10)8-7-11(2)14/h6-8,12H,5,9H2,1-4H3/b8-7+

InChI key

UZFLPKAIBPNNCA-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

244.4 °F - closed cup

Flash Point(C)

118 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yonghong Wu et al.
Environmental microbiology, 13(3), 604-615 (2010-11-09)
Periphyton biofilms are natural mixtures comprised of photoautotrophic and heterotrophic complex microorganisms. In this work, the inhibition effects of periphyton biofilms on cyanobacterial blooms were studied in pilot and field trials. Results show that the cyanobacterial species responsible for the
J Lalko et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 45 Suppl 1, S251-S257 (2007-11-23)
A toxicologic and dermatologic review of ionone when used as a fragrance ingredient is presented.
Sabine Sewenig et al.
Journal of agricultural and food chemistry, 53(4), 838-844 (2005-02-17)
A new coupling system of GC-GC, connected via a Multi Column Switching Device MCS2 for measuring isotope ratios, is introduced. By means of several standard substances the precise and accurate measurement of isotopic values is proved. First applications concerning the
Ufuk Ozgen et al.
Molecules (Basel, Switzerland), 15(4), 2593-2599 (2010-04-30)
Sonchus erzincanicus (Asteraceae) is an endemic species in Turkey, where six Sonchus species grow. In this study, a phytochemical study was performed on the aerial parts of the plant. The study describes the isolation and structure elucidation of five flavonoids
Marco Luparia et al.
Chemistry & biodiversity, 5(6), 1045-1057 (2008-07-12)
To study the influence of the steric bulk of the substituents at C(5) on the olfactory characteristics of alpha-ionone, the (S)-antipodes of compounds 8-10 were synthesized starting from (S)-alpha-cyclogeraniol (14a). The latter was available in useful preparative yield with 95%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service