Skip to Content
Merck
All Photos(2)

Key Documents

H46007

Sigma-Aldrich

3-Hydroxy-2-naphthoic acid

98%

Synonym(s):

β-Hydroxynaphthoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC10H6CO2H
CAS Number:
Molecular Weight:
188.18
Beilstein:
744100
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

mp

218-221 °C (lit.)

SMILES string

OC(=O)c1cc2ccccc2cc1O

InChI

1S/C11H8O3/c12-10-6-8-4-2-1-3-7(8)5-9(10)11(13)14/h1-6,12H,(H,13,14)

InChI key

ALKYHXVLJMQRLQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philip Pirkwieser et al.
Frontiers in chemistry, 6, 172-172 (2018-06-08)
Ionic liquids (ILs) are per definition salts with melting points below 100°C and might be green alternatives for the extraction of heavy metals from aqueous solutions due to their favorable environmental and physico-chemical properties. Partial solution during extraction, so-called leaching
Pinki Saha Sardar et al.
The journal of physical chemistry. B, 112(11), 3451-3461 (2008-02-26)
The steady-state and time-resolved studies of the sensitized emission of the excited-state proton transfer (ESIPT) probe 3-hydroxy-2-naphthoic acid (3HNA) when bound to bovine serum albumin (BSA) and human serum albumin (HSA) indicate that the nonradiative dipole-dipole Förster type energy transfer
F Cañada-Cañada et al.
Journal of fluorescence, 17(1), 23-28 (2006-12-13)
A spectrofluorimetric method has been developed for the determination of 3-hydroxy-2-naphthoic acid (3H2NA) by formation of a ternary complex with zirconium (IV) and beta-cyclodextrin (beta-CD). It has been observed that the fluorescence intensity of 3H2NA is greatly enhanced when the
Marianita Perez-Gonzalez et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 36(2), 212-224 (2019-01-31)
Pigment Red 57 (Colour Index No. 15850, PR57) and Pigment Red 57:1 (Colour Index No. 15850:1, PR57:1) are certifiable in the USA as the color additives D&C Red No. 6 (R6) and D&C Red No. 7 (R7) for use in
Jason R Smith et al.
Bioorganic & medicinal chemistry, 20(3), 1354-1363 (2011-11-25)
Indoleamine 2,3-dioxygenase-1 (IDO-1) is a heme containing enzyme that catalyses the initial step in the major pathway of l-tryptophan catabolism; the kynurenine pathway. A large body of evidence has been accumulating for its immunosuppressive and tumoural escape roles and its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service