Skip to Content
Merck
All Photos(2)

Key Documents

H2131

Sigma-Aldrich

1-Hexadecene

≥98.5%

Synonym(s):

Cetene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13CH=CH2
CAS Number:
Molecular Weight:
224.43
Beilstein:
1753401
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

>0.8 (vs air)

vapor pressure

<0.2 mmHg ( 20 °C)

Assay

≥98.5%

form

liquid

autoignition temp.

464 °F

refractive index

n20/D 1.441 (lit.)

bp

274 °C (lit.)

mp

3-5 °C (lit.)

density

0.783 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCC=C

InChI

1S/C16H32/c1-3-5-7-9-11-13-15-16-14-12-10-8-6-4-2/h3H,1,4-16H2,2H3

InChI key

GQEZCXVZFLOKMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

1-Hexadecene, a long-chain alpha-olefin (LAO)containing 16 carbon atoms, commonly used as a precursor in the production ofsynthetic detergents, plasticizers, and various chemicals for the chemicalindustry. Additionally, it is a key component in commercial LAO blends,contributing to the manufacturing of tanning oils, synthetic fatty acids, anddrilling fluids.[1] [2]

Application

  • Development of petroleum lubricating oil additives: A study evaluated newly synthesized copolymers and their montmorillonite nanocomposites as cold flow improvers for petroleum lubricating oils, including formulations with 1-Hexadecene, demonstrating enhanced flow properties (El-Bahnasawi et al., 2023).
  • Attractants for biocontrol agents: Research identified volatile blends containing 1-Hexadecene that attract the biocontrol agent Galerucella placida, suggesting applications in agricultural pest management (Koner et al., 2022).
  • Adsorbent materials for oil spill cleanup: A study explored the modification of commercial sponges with 1-Hexadecene-based polymers for efficient adsorption of crude oil and organic solvents, highlighting potential environmental cleanup applications (Jin et al., 2022).
  • Surface engineering for hydrophobicity: 1-Hexadecene was used to create organosilane micropatterns on surfaces, improving their hydrophobic properties, relevant in material science and surface engineering (Soliman et al., 2021).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Genome and metabolome analysis of Bacillus sp. Hex-HIT36: A newly screened functional microorganism for the degradation of 1-hexadecene in industrial wastewater
Yang, Lei and Chen et,al.
Environmental Research, 260, 119594-119594 (2024)
Enhancement of methane production from 1-hexadecene by additional electron donors
Paulo, AMS and Salvador, et, al.
Microbial Biotechnology, 11, 657-666 (2018)
Contact allergy to polyvinylpyrrolidone/hexadecene copolymer previously reported.
A C De Groot
American journal of contact dermatitis : official journal of the American Contact Dermatitis Society, 7(4), 260-260 (1996-12-01)
M Tobler et al.
Contact dermatitis, 26(5), 299-303 (1992-05-01)
In continuation of our preceding studies, we disclose long-term experiments to test more extensively the protective power of the novel 4H Glove. 2 compounds encountered in modern electron microscopy technique, 1-hexadecene (1-HD) and 2-hydroxyethyl acrylate (2-HEA), which elicit allergic and/or
Yasuyuki Kubo et al.
mBio, 6(5), e01305-e01315 (2015-09-17)
The cucumber anthracnose fungus Colletotrichum orbiculare forms specialized cells called appressoria for host penetration. We identified a gene, FAM1, encoding a novel peroxin protein that is essential for peroxisome biogenesis and that associates with Woronin bodies (WBs), dense-core vesicles found

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service