GF98785388
Vanadium
microfoil, disks, 10mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Synonym(s):
Vanadium, V 004500
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
vapor pressure
8 mmHg ( 20 °C)
Assay
≥99.8%
manufacturer/tradename
Goodfellow 987-853-88
resistivity
24.8-26.0 μΩ-cm, 20°C
bp
3380 °C (lit.)
mp
1890 °C (lit.)
density
6.11 g/mL at 25 °C (lit.)
SMILES string
[V]
InChI
1S/V
InChI key
LEONUFNNVUYDNQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Cell biochemistry and biophysics, 44(1), 73-81 (2006-02-04)
Among several metals, vanadium has emerged as an extremely potent agent with insulin-like properties. These insulin-like properties have been demonstrated in isolated cells, tissues, different animal models of type I and type II diabetes as well as a limited number
Journal of inorganic biochemistry, 102(5-6), 1152-1158 (2008-02-08)
In the present focused review, vanadate-dependent haloperoxidases and vanadate-inhibited enzymes which catalyze the hydrolysis of phosphoester bonds are addressed. In these systems, vanadate [HxVO4](3-x)(-) is covalently coordinated to the imidazolyl moiety of an active site histidine, with a geometrical arrangement
ChemSusChem, 4(10), 1388-1406 (2011-11-22)
Large-scale energy storage has become the main bottleneck for increasing the percentage of renewable energy in our electricity grids. Redox flow batteries are considered to be among the best options for electricity storage in the megawatt range and large demonstration
Canadian journal of physiology and pharmacology, 82(10), 833-839 (2004-12-02)
Inorganic and organic compounds of vanadium have been shown to exhibit a large range of insulinomimetic effects in the cardiovascular system, including stimulation of glucose transporter 4 (GLUT-4) translocation and glucose transport in adult cardiomyocytes. Furthermore, administration of vanadium compounds
Journal of hazardous materials, 296, 158-165 (2015-04-29)
Blast furnace (BF) slags are commonly applied as soil amendments and in road fill material. In Sweden they are also naturally high in vanadium. The aim of this study was to assess the vanadium bioavailability in BF slags when applied
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service