Skip to Content
Merck
All Photos(2)

Key Documents

GF96440676

Chromium

foil, not light tested, 50x50mm, thickness 0.03mm, permanent polyester support, 99.99+%

Synonym(s):

Chromium, CR000170

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cr
CAS Number:
Molecular Weight:
52.00
MDL number:
UNSPSC Code:
12141709
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.99%

form

foil

manufacturer/tradename

Goodfellow 964-406-76

resistivity

12.7 μΩ-cm, 20°C

bp

2672 °C (lit.)

mp

1857 °C (lit.)

density

7.14 g/mL at 25 °C (lit.)

SMILES string

[Cr]

InChI

1S/Cr

InChI key

VYZAMTAEIAYCRO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra S Wise et al.
Mutation research, 733(1-2), 78-82 (2011-12-24)
Many metals serve as micronutrients which protect against genomic instability. Chromium is most abundant in its trivalent and hexavalent forms. Trivalent chromium has historically been considered an essential element, though recent data indicate that while it can have pharmacological effects
Akiko Maehara et al.
EuroIntervention : journal of EuroPCR in collaboration with the Working Group on Interventional Cardiology of the European Society of Cardiology, 10(4), 475-483 (2014-08-21)
To report serial intravascular ultrasound (IVUS) findings of bifurcation lesions treated with the dedicated Tryton Side Branch Stent to assess mechanisms of restenosis. The Tryton FIM study was a multicentre, prospective, single-arm, "first-in-man" study to treat de novo bifurcation lesions.
B Dhal et al.
Journal of hazardous materials, 250-251, 272-291 (2013-03-08)
Chromium is a highly toxic non-essential metal for microorganisms and plants, and its occurrence is rare in nature. Lower to higher chromium containing effluents and solid wastes released by activities such as mining, metal plating, wood preservation, ink manufacture, dyes
A M Urbano et al.
Current drug metabolism, 13(3), 284-305 (2012-03-30)
For over a century, chromium (Cr) has found widespread industrial and commercial use, namely as a pigment, in the production of stainless steel and in chrome plating. The adverse health effects to the skin and respiratory tract of prolonged exposure
Satyendra Kumar Garg et al.
Reviews of environmental contamination and toxicology, 217, 75-140 (2012-02-22)
Bioremediation offers the possibility of using living organisms (bacteria, fungi, algae,or plants), but primarily microorganisms, to degrade or remove environmental contaminants, and transform them into nontoxic or less-toxic forms. The major advantages of bioremediation over conventional physicochemical and biological treatment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service