Skip to Content
Merck
All Photos(2)

Key Documents

GF94328387

Tin

rod, 200mm, diameter 5.0mm, extruded, 99.75%

Synonym(s):

Tin, SN007920

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sn
CAS Number:
Molecular Weight:
118.71
MDL number:
UNSPSC Code:
12141745
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.75%

form

rod

manufacturer/tradename

Goodfellow 943-283-87

resistivity

11 μΩ-cm, 20°C

bp

2270 °C (lit.)

mp

231.9 °C (lit.)

density

7.310 g/mL at 25 °C (lit.)

SMILES string

[Sn]

InChI

1S/Sn

InChI key

ATJFFYVFTNAWJD-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The newer essential trace elements, chromium, tin, nickel, vanadium and silicon.
W Mertz
The Proceedings of the Nutrition Society, 33(3), 307-313 (1974-12-01)
[Tin in the environment].
Iu P Popov et al.
Gigiena i sanitariia, (9)(9), 55-57 (1983-09-01)
Inorganic tin: chemistry, disposition and role in nuclear medicine diagnostic skeletal imaging agents.
M D Francis et al.
International journal of nuclear medicine and biology, 8(2-3), 145-152 (1981-01-01)
K A Winship
Adverse drug reactions and acute poisoning reviews, 7(1), 19-38 (1988-01-01)
Inorganic tin salts are poorly absorbed and rapidly excreted in the faeces; as a result they have a low toxicity. Only about 5 per cent is absorbed from the gastrointestinal tract, widely distributed in the body, then excreted by the
F J de Araujo et al.
Genes and immunity, 16(4), 284-288 (2015-03-13)
Functional variations in the mannose-binding lectin (MBL2) gene causing low levels of serum MBL are associated with susceptibility to numerous infectious diseases. We investigated whether there is genetic association of MBL2 variant alleles with cutaneous leishmaniasis (CL) caused by Leishmania

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service