GF83108695
Nickel-manganese alloy, Ni75Mn25
powder, 45micron max. particle size, atomized, 200g
Synonym(s):
NI256045
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
powder
manufacturer/tradename
Goodfellow 831-086-95
weight
200 g
max. part. size
45 μm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service