GF73574231
Chromium
microfoil, disks, 10mm, thinness 0.05μm, specific density 37.27μg/cm2, permanent mylar 3.5μm support, 99.99+%
Synonym(s):
Chromium, CR004300
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Assay
≥99.99%
form
foil
manufacturer/tradename
Goodfellow 735-742-31
resistivity
12.7 μΩ-cm, 20°C
bp
2672 °C (lit.)
mp
1857 °C (lit.)
density
7.14 g/mL at 25 °C (lit.)
SMILES string
[Cr]
InChI
1S/Cr
InChI key
VYZAMTAEIAYCRO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Current drug metabolism, 13(3), 284-305 (2012-03-30)
For over a century, chromium (Cr) has found widespread industrial and commercial use, namely as a pigment, in the production of stainless steel and in chrome plating. The adverse health effects to the skin and respiratory tract of prolonged exposure
Mutation research, 733(1-2), 78-82 (2011-12-24)
Many metals serve as micronutrients which protect against genomic instability. Chromium is most abundant in its trivalent and hexavalent forms. Trivalent chromium has historically been considered an essential element, though recent data indicate that while it can have pharmacological effects
EuroIntervention : journal of EuroPCR in collaboration with the Working Group on Interventional Cardiology of the European Society of Cardiology, 10(4), 475-483 (2014-08-21)
To report serial intravascular ultrasound (IVUS) findings of bifurcation lesions treated with the dedicated Tryton Side Branch Stent to assess mechanisms of restenosis. The Tryton FIM study was a multicentre, prospective, single-arm, "first-in-man" study to treat de novo bifurcation lesions.
Journal of hazardous materials, 250-251, 272-291 (2013-03-08)
Chromium is a highly toxic non-essential metal for microorganisms and plants, and its occurrence is rare in nature. Lower to higher chromium containing effluents and solid wastes released by activities such as mining, metal plating, wood preservation, ink manufacture, dyes
Reviews of environmental contamination and toxicology, 217, 75-140 (2012-02-22)
Bioremediation offers the possibility of using living organisms (bacteria, fungi, algae,or plants), but primarily microorganisms, to degrade or remove environmental contaminants, and transform them into nontoxic or less-toxic forms. The major advantages of bioremediation over conventional physicochemical and biological treatment
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service