Skip to Content
Merck
All Photos(2)

Key Documents

GF71274258

Manganese

foil, not light tested, 50x50mm, thickness 0.008mm, permanent polyester support, 98.7%

Synonym(s):

Manganese, MN000130

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mn
CAS Number:
Molecular Weight:
54.94
MDL number:
UNSPSC Code:
12141724
PubChem Substance ID:
NACRES:
NA.23

Assay

98.7%

form

foil

manufacturer/tradename

Goodfellow 712-742-58

resistivity

185 μΩ-cm, 20°C

size × thickness

50x50 mm × 0.008 mm

bp

1962 °C (lit.)

mp

1244 °C (lit.)

density

7.3 g/mL at 25 °C (lit.)

SMILES string

[Mn]

InChI

1S/Mn

InChI key

PWHULOQIROXLJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mirna Lechpammer et al.
Annals of neurology, 75(4), 608-612 (2014-03-07)
We followed a patient with manganese transporter deficiency due to homozygous SLC30A10 mutations from age 14 years until his death at age 38 years and present the first postmortem findings of this disorder. The basal ganglia showed neuronal loss, rhodanine-positive
Srinivasan V Narayanan et al.
Stroke, 46(6), 1626-1632 (2015-04-25)
Nuclear erythroid 2 related factor 2 (Nrf2) is an astrocyte-enriched transcription factor that has previously been shown to upregulate cellular antioxidant systems in response to ischemia. Although resveratrol preconditioning (RPC) has emerged as a potential neuroprotective therapy, the involvement of
Mohammad Mahdi Najafpour et al.
Photosynthesis research, 114(1), 1-13 (2012-09-04)
The resolution of Photosystem II (PS II) crystals has been improved using isolated PS II from the thermophilic cyanobacterium Thermosynechococcus vulcanus. The new 1.9 Å resolution data have provided detailed information on the structure of the water-oxidizing complex (Umena et
Ebany J Martinez-Finley et al.
Free radical biology & medicine, 62, 65-75 (2013-02-12)
Manganese (Mn) is an essential dietary nutrient, but an excess or accumulation can be toxic. Disease states, such as manganism, are associated with overexposure or accumulation of Mn and are due to the production of reactive oxygen species, free radicals
Karin Tuschl et al.
International review of neurobiology, 110, 277-312 (2013-11-12)
Manganese (Mn) is an essential trace metal that is pivotal for normal cell function and metabolism. Its homeostasis is tightly regulated; however, the mechanisms of Mn homeostasis are poorly characterized. While a number of proteins such as the divalent metal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service