Skip to Content
Merck
All Photos(2)

Key Documents

GF71248013

Nickel

mesh, size 0.43 mm, size 60 × 120 mm, thickness 0.004 mm

Synonym(s):

NI008720

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
MDL number:
UNSPSC Code:
12141729
PubChem Substance ID:
NACRES:
NA.23

form

mesh

manufacturer/tradename

Goodfellow 712-480-13

resistivity

6.97 μΩ-cm, 20°C

size

0.43 mm
60 × 120 mm

thickness

0.004 mm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Srinidhi Nagaraja et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 108(6), 2681-2690 (2020-03-12)
Nitinol is commonly used in medical implants due to its unique thermomechanical properties of shape memory and superelasticity. Free nickel has the potential to induce biological responses that may be a concern for permanent implants manufactured from nickel-containing alloys. Although

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service