Skip to Content
Merck
All Photos(2)

Key Documents

GF48730541

Tungsten

wire, 0.457 mm diameter, size 300 mm, condition straight wire

Synonym(s):

W 005457, W

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€152.90
250 MG
€198.40
500 MG
€286.80
1 G
€354.40
2.5 G
€572.80
5 G
€936.80
10 G
€1,664.80

Vendor SKU: EN300-127045-100MG

€152.90


Ships in 14 days - from Aldrich Partner.

To order products, please contact your local dealer.

Select a Size

Change View
100 MG
€152.90
250 MG
€198.40
500 MG
€286.80
1 G
€354.40
2.5 G
€572.80
5 G
€936.80
10 G
€1,664.80

About This Item

Empirical Formula (Hill Notation):
W
CAS Number:
Molecular Weight:
183.84
MDL number:
UNSPSC Code:
12141747
PubChem Substance ID:
NACRES:
NA.23

Vendor SKU: EN300-127045-100MG

€152.90


Ships in 14 days - from Aldrich Partner.

To order products, please contact your local dealer.

form

wire

manufacturer/tradename

Goodfellow 487-305-41

resistivity

4.9 μΩ-cm, 20°C

size

300 mm

diameter

0.457 mm

bp

5660 °C (lit.)

mp

3410 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

SMILES string

[W]

InChI

1S/W

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service