Skip to Content
Merck
All Photos(2)

Key Documents

GF43284416

Chromium

microfoil, disks, 10mm, thinness 0.5μm, specific density 355μg/cm2, permanent mylar 3.5μm support, 99.99+%

Synonym(s):

Chromium, CR004700

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cr
CAS Number:
Molecular Weight:
52.00
MDL number:
UNSPSC Code:
12141709
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.99%

form

foil

manufacturer/tradename

Goodfellow 432-844-16

resistivity

12.7 μΩ-cm, 20°C

L × thickness

10 mm × 0.5 μm

bp

2672 °C (lit.)

mp

1857 °C (lit.)

density

7.14 g/mL at 25 °C (lit.)

SMILES string

[Cr]

InChI

1S/Cr

InChI key

VYZAMTAEIAYCRO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satyendra Kumar Garg et al.
Reviews of environmental contamination and toxicology, 217, 75-140 (2012-02-22)
Bioremediation offers the possibility of using living organisms (bacteria, fungi, algae,or plants), but primarily microorganisms, to degrade or remove environmental contaminants, and transform them into nontoxic or less-toxic forms. The major advantages of bioremediation over conventional physicochemical and biological treatment
Chad M Thompson et al.
Critical reviews in toxicology, 43(3), 244-274 (2013-03-01)
Abstract Chronic exposure to high concentrations of hexavalent chromium (Cr(VI)) in drinking water causes intestinal adenomas and carcinomas in mice, but not in rats. Cr(VI) causes damage to intestinal villi and crypt hyperplasia in mice after only one week of
Yinan Hua et al.
The Journal of nutritional biochemistry, 23(4), 313-319 (2012-03-20)
Type 2 diabetes is often associated with obesity, dyslipidemia and cardiovascular anomalies and is a major health problem approaching global epidemic proportions. Insulin resistance, a prediabetic condition, precedes the onset of frank type 2 diabetes and offers potential avenues for
Erik Wennerberg et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(22), 5733-5744 (2014-09-13)
Anaplastic thyroid carcinoma (ATC) is one of the most aggressive forms of cancer with no curative therapies available. To date, strategies to target ATC by immunotherapy have not been evaluated. We investigated whether ATC would be a suitable target for
A Peña-Fernández et al.
Environment international, 72, 176-185 (2014-05-06)
Rapid development, industrialisation, and urbanisation have resulted in serious contamination of soil by metals and metalloids from anthropogenic sources in many areas of the world, either directly or indirectly. Exponential urban and economic development has resulted in human populations settling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service