Skip to Content
Merck
All Photos(2)

Key Documents

GF31001960

Bismuth

foil, 25x25mm, thickness 2.0mm, 100%

Synonym(s):

Bismuth, BI000300

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Bi
CAS Number:
Molecular Weight:
208.98
MDL number:
UNSPSC Code:
12141706
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

100%

form

foil

manufacturer/tradename

Goodfellow 310-019-60

resistivity

129 μΩ-cm, 20°C

size × thickness

25x25 mm × 2.0 mm

bp

1560 °C (lit.)

mp

271 °C (lit.)

density

9.8 g/mL at 25 °C (lit.)

SMILES string

[Bi]

InChI

1S/Bi

InChI key

JCXGWMGPZLAOME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Javier P Gisbert et al.
Helicobacter, 10(3), 157-171 (2005-05-21)
To systematically review the Helicobacter pylori eradication efficacy with ranitidine bismuth citrate (RBC) and two antibiotics, and to conduct a meta-analysis of randomized clinical trials comparing the efficacy of proton pump inhibitor (PPI) vs. RBC with two antibiotics for 1
Montserrat Filella
Journal of environmental monitoring : JEM, 12(1), 90-109 (2010-01-19)
Like all elements of the periodic table, bismuth is ubiquitously distributed throughout the environment as a result of natural processes and human activities. It is present as Bi(III) in environmental, biological and geochemical samples. Although bismuth and its compounds are
Emma Aneheim et al.
Scientific reports, 5, 12025-12025 (2015-07-15)
To facilitate multicentre clinical studies on targeted alpha therapy, it is necessary to develop an automated, on-site procedure for conjugating rare, short-lived, alpha-emitting radionuclides to biomolecules. Astatine-211 is one of the few alpha-emitting nuclides with appropriate chemical and physical properties
Bismuth in medicine.
Hongzhe Sun et al.
Metal ions in biological systems, 41, 333-378 (2004-06-23)
Hong Lu et al.
European journal of gastroenterology & hepatology, 25(10), 1134-1140 (2013-06-20)
Antimicrobial resistance has continued to undermine many popular anti-Helicobacter pylori therapies. Antibiotic resistance to commonly used anti-H. pylori drugs in China has increased markedly, making China an ideal site to identify regimens that remain effective despite widespread antimicrobial resistance. Bismuth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service