Skip to Content
Merck
All Photos(2)

Key Documents

GF21606386

Erbium

rod, 50mm, diameter 10.0mm, cast, 99%

Synonym(s):

Erbium, ER007920

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Er
CAS Number:
Molecular Weight:
167.26
MDL number:
UNSPSC Code:
12141603
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

rod

manufacturer/tradename

Goodfellow 216-063-86

resistivity

86 μΩ-cm, 20°C

L × diam.

50 mm × 10.0 mm

bp

2868 °C (lit.)

mp

1529 °C (lit.)

density

9.062 g/mL at 25 °C (lit.)

SMILES string

[Er]

InChI

1S/Er

InChI key

UYAHIZSMUZPPFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isao Ishikawa et al.
Journal of periodontal research, 39(4), 275-285 (2004-06-23)
Since lasers were introduced for the treatment of oral diseases, there has been considerable advancement in technology. As a result, numerous laser systems are currently available for oral use. Neodymium:Yttrium-Aluminum:Garnet (Nd:YAG), carbon dioxide (CO(2)) laser and the semiconductor Diode lasers
Francesco Enrichi
Annals of the New York Academy of Sciences, 1130, 262-266 (2008-07-04)
This work presents the morphological and optical properties of luminescent silica spheres, discussing applications in bioimaging and biosensing. The spheres are obtained by the hydrolysis and condensation of tetraethylorthosilicate (TEOS) and can be synthesized by following either a basic or
Glenn van As
Dental clinics of North America, 48(4), 1017-1059 (2004-10-07)
Erbium hard tissue lasers have the capability to prepare enamel, dentin, caries, cementum, and bone in addition to cutting soft tissue. The ability of hard tissue lasers to reduce or eliminate vibrations, the audible whine of drills, microfractures, and some
David M Bowman et al.
Journal of long-term effects of medical implants, 13(6), 503-508 (2004-04-02)
Erbium:yttrium-aluminum-garnet (Er:YAG) cataract extraction has shown considerable promise as a new technique for lens nucleus removal prior to intraocular lens (IOL) implantation for the treatment of cataracts. Recent studies suggest that the laser energy is dissipated much more locally and
T S Alster et al.
Dermatologic clinics, 19(3), 453-466 (2001-10-16)
The short-pulsed Er:YAG laser system is an excellent ablative tool for cutaneous resurfacing. This system is most efficacious for patients with milder cutaneous involvement, including mild photoinduced facial rhytides, mildly atrophic scars, and textural changes caused by fibrosis and dermatochalasis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service